Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trifluorophenylboronic acid, 95+%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
143418-49-9
C6H4BF3O2
175.901
MFCD02093069
UHDDEIOYXFXNNJ-UHFFFAOYSA-N
3,4,5-trifluorophenyl boronic acid,3,4,5-trifluorobenzeneboronic acid,3,4,5-trifluorophenyl boranediol,boronic acid, b-3,4,5-trifluorophenyl,3,4,5-trifluoro phenylboric acid,boronic acid, 3,4,5-trifluorophenyl,3,4,5-trifluorophenylboronicacid,pubchem2340,acmc-209cqj
2734671
(3,4,5-trifluorophenyl)boronic acid
B(C1=CC(=C(C(=C1)F)F)F)(O)O
143418-49-9
C6H4BF3O2
175.901
MFCD02093069
UHDDEIOYXFXNNJ-UHFFFAOYSA-N
3,4,5-trifluorophenyl boronic acid,3,4,5-trifluorobenzeneboronic acid,3,4,5-trifluorophenyl boranediol,boronic acid, b-3,4,5-trifluorophenyl,3,4,5-trifluoro phenylboric acid,boronic acid, 3,4,5-trifluorophenyl,3,4,5-trifluorophenylboronicacid,pubchem2340,acmc-209cqj
2734671
(3,4,5-trifluorophenyl)boronic acid
B(C1=CC(=C(C(=C1)F)F)F)(O)O
Color Tan
Melting Point 284.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95+%
Formula Weight 175.9
Physical Form Powder
Percent Purity ≥95%
Chemical Name or Material 3,4,5-Trifluorophenylboronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning