Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxyaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
24313-88-0
C9H13NO3
183.21
MFCD00008393
XEFRNCLPPFDWAC-UHFFFAOYSA-N
benzenamine, 3,4,5-trimethoxy,3,4,5-trimethoxy aniline,3,4,5-trimethoxybenzenamine,3,4,5-trimethoxy-phenylamine,3,4,5-trimethoxyphenylamine,3,5-trimethoxyaniline,3,4,5-trimethoxyanilin,acmc-1cdy7,intermediates-zcf02654,3,4,5-trimethoxyaniline
32285
3,4,5-trimethoxyaniline
COC1=CC(=CC(=C1OC)OC)N
24313-88-0
C9H13NO3
183.21
MFCD00008393
XEFRNCLPPFDWAC-UHFFFAOYSA-N
benzenamine, 3,4,5-trimethoxy,3,4,5-trimethoxy aniline,3,4,5-trimethoxybenzenamine,3,4,5-trimethoxy-phenylamine,3,4,5-trimethoxyphenylamine,3,5-trimethoxyaniline,3,4,5-trimethoxyanilin,acmc-1cdy7,intermediates-zcf02654,3,4,5-trimethoxyaniline
32285
3,4,5-trimethoxyaniline
COC1=CC(=CC(=C1OC)OC)N
Infrared Spectrum Authentic
Melting Point 110.0°C to 113.0°C
Color Pink
Quantity 25g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3O)3C6H2NH2
Beilstein 13, 826
Solubility Information Solubility in water: insoluble
Formula Weight 183.21
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 3, 4, 5-Trimethoxyaniline
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning