Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxyaniline 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
24313-88-0
C9H13NO3
183.207
MFCD00008393
XEFRNCLPPFDWAC-UHFFFAOYSA-N
benzenamine, 3,4,5-trimethoxy,3,4,5-trimethoxy aniline,3,4,5-trimethoxybenzenamine,3,4,5-trimethoxy-phenylamine,3,4,5-trimethoxyphenylamine,3,5-trimethoxyaniline,3,4,5-trimethoxyanilin,acmc-1cdy7,intermediates-zcf02654,3,4,5-trimethoxyaniline
32285
3,4,5-trimethoxyaniline
COC1=CC(=CC(=C1OC)OC)N
24313-88-0
C9H13NO3
183.207
MFCD00008393
XEFRNCLPPFDWAC-UHFFFAOYSA-N
benzenamine, 3,4,5-trimethoxy,3,4,5-trimethoxy aniline,3,4,5-trimethoxybenzenamine,3,4,5-trimethoxy-phenylamine,3,4,5-trimethoxyphenylamine,3,5-trimethoxyaniline,3,4,5-trimethoxyanilin,acmc-1cdy7,intermediates-zcf02654,3,4,5-trimethoxyaniline
32285
3,4,5-trimethoxyaniline
COC1=CC(=CC(=C1OC)OC)N
Color Undesignated
Melting Point 112°C
Quantity 25g
Formula Weight 183.21
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3,4,5-Trimethoxyaniline