Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxybenzaldehyde, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
86-81-7
C10H12O4
196.2
MFCD00003364
OPHQOIGEOHXOGX-UHFFFAOYSA-N
benzaldehyde, 3,4,5-trimethoxy,unii-wl86yd76n6,tmba,benzaldehyde,3,4,5-trimethoxy,3,4,5-trimethoxy benzaldehyde,3,4,5-trimethoxy-benzaldehyde,pubchem8266,acmc-209qbj,3,5-trimethoxybenzaldehyde,2.3.4-trihyroxybenzaldehyde
6858
3,4,5-trimethoxybenzaldehyde
COC1=CC(=CC(=C1OC)OC)C=O
86-81-7
C10H12O4
196.2
MFCD00003364
OPHQOIGEOHXOGX-UHFFFAOYSA-N
benzaldehyde, 3,4,5-trimethoxy,unii-wl86yd76n6,tmba,benzaldehyde,3,4,5-trimethoxy,3,4,5-trimethoxy benzaldehyde,3,4,5-trimethoxy-benzaldehyde,pubchem8266,acmc-209qbj,3,5-trimethoxybenzaldehyde,2.3.4-trihyroxybenzaldehyde
6858
3,4,5-trimethoxybenzaldehyde
COC1=CC(=CC(=C1OC)OC)C=O
Melting Point 73.0°C to 76.0°C
Infrared Spectrum Authentic
Quantity 500g
Packaging Glass bottle
Color Cream to White
Boiling Point 163.0°C to 165.0°C (10.0mmHg)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3O)3C6H2CHO
Beilstein 08, 391
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,toluene and acetone
Formula Weight 196.2
Physical Form Powder or Flakes
Percent Purity 99%
Chemical Name or Material 3, 4, 5-Trimethoxybenzaldehyde
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning