Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxybenzoic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
118-41-2
C10H12O5
212.201
MFCD00002501
SJSOFNCYXJUNBT-UHFFFAOYSA-N
eudesmic acid,gallic acid trimethyl ether,tri-o-methylgallic acid,trimethylgallic acid,benzoic acid, 3,4,5-trimethoxy,veratric acid, 5-methoxy,unii-v5c9h0sc9f,3,4,5-trimethoxy-benzoic acid,5-methoxy-veratric acid,v5c9h0sc9f
8357
CHEBI:454991
3,4,5-trimethoxybenzoic acid
COC1=CC(=CC(=C1OC)OC)C(=O)O
118-41-2
C10H12O5
212.201
MFCD00002501
SJSOFNCYXJUNBT-UHFFFAOYSA-N
eudesmic acid,gallic acid trimethyl ether,tri-o-methylgallic acid,trimethylgallic acid,benzoic acid, 3,4,5-trimethoxy,veratric acid, 5-methoxy,unii-v5c9h0sc9f,3,4,5-trimethoxy-benzoic acid,5-methoxy-veratric acid,v5c9h0sc9f
8357
CHEBI:454991
3,4,5-trimethoxybenzoic acid
COC1=CC(=CC(=C1OC)OC)C(=O)O
Packaging Glass Bottle
Boiling Point 225.0°C to 227.0°C (10.0mmHg)
Quantity 5g
Infrared Spectrum Authentic
Color Colorless to Yellow
Melting Point 168.0°C to 172.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3O)3C6H2CO2H
Beilstein 10, 481
Formula Weight 212.2
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3, 4, 5-Trimethoxybenzoic acid, 99%
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber : 204-248-2

TSCA : TSCA