Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxybenzylamine, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
18638-99-8
C10H15NO3
197.24
MFCD00017150
YUPUSBMJCFBHAP-UHFFFAOYSA-N
3,4,5-trimethoxybenzylamine,3,4,5-trimethoxyphenyl methanamine,1-3,4,5-trimethoxyphenyl methanamine,benzenemethanamine, 3,4,5-trimethoxy,3,4,5-trimethoxyphenyl methylamine,1-3,4,5-trimethoxyphenyl methanamine hydrochloride,pubchem16674,acmc-1c5ez,ksc495e6b,3,4,5-trimethoxy benzylamine
87736
(3,4,5-trimethoxyphenyl)methanamine
COC1=CC(=CC(=C1OC)OC)CN
18638-99-8
C10H15NO3
197.24
MFCD00017150
YUPUSBMJCFBHAP-UHFFFAOYSA-N
3,4,5-trimethoxybenzylamine,3,4,5-trimethoxyphenyl methanamine,1-3,4,5-trimethoxyphenyl methanamine,benzenemethanamine, 3,4,5-trimethoxy,3,4,5-trimethoxyphenyl methylamine,1-3,4,5-trimethoxyphenyl methanamine hydrochloride,pubchem16674,acmc-1c5ez,ksc495e6b,3,4,5-trimethoxy benzylamine
87736
(3,4,5-trimethoxyphenyl)methanamine
COC1=CC(=CC(=C1OC)OC)CN
Specific Gravity 1.155
Refractive Index 1.5455 to 1.5485
Packaging Glass bottle
Quantity 5g
Density 1.1550g/mL
Flash Point >110°C
Color Colorless to Yellow
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula (CH3O)3C6H2CH2NH2
Formula Weight 197.24
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 3, 4, 5-Trimethoxybenzylamine, 96%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 242-468-0

TSCA : TSCA