Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxyphenol, 98.5+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
642-71-7
C9H12O4
184.19
MFCD00008389
VTCDZPUMZAZMSB-UHFFFAOYSA-N
antiarol,phenol, 3,4,5-trimethoxy,3,4,5-trimethoxy phenol,spectrum_000525,specplus_000623,spectrum3_001208,spectrum4_001487,spectrum5_000193,3,4,5-trirnethoxyphenol,3,4,5-trimethoxy-phenol
69505
CHEBI:2760
3,4,5-trimethoxyphenol
COC1=CC(=CC(=C1OC)OC)O
642-71-7
C9H12O4
184.19
MFCD00008389
VTCDZPUMZAZMSB-UHFFFAOYSA-N
antiarol,phenol, 3,4,5-trimethoxy,3,4,5-trimethoxy phenol,spectrum_000525,specplus_000623,spectrum3_001208,spectrum4_001487,spectrum5_000193,3,4,5-trirnethoxyphenol,3,4,5-trimethoxy-phenol
69505
CHEBI:2760
3,4,5-trimethoxyphenol
COC1=CC(=CC(=C1OC)OC)O
Melting Point 146.0°C to 149.0°C
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color Brown to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula (CH3O)3C6H2OH
Beilstein 06,1154
Formula Weight 184.19
Physical Form Powder and/or Chunks
Percent Purity 98.5+%
Chemical Name or Material 3, 4, 5-Trimethoxyphenol, 98.5+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-387-2

TSCA : TSCA