Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxyphenylacetic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
Packaging:
Glass bottle
951-82-6
C11H14O5
226.23
MFCD00004336
DDSJXCGGOXKGSJ-UHFFFAOYSA-N
3,4,5-trimethoxyphenylacetic acid,2-3,4,5-trimethoxyphenyl acetic acid,3,4,5-trimethoxyphenyl acetic acid,benzeneacetic acid, 3,4,5-trimethoxy,3,4,5-trimethoxyphenylaceticacid,pubchem14015,acmc-209rv8,ksc487g4j,3,4,5-trimethoxybenzeneacetic acid,benzeneacetic acid,4,5-trimethoxy
70372
2-(3,4,5-trimethoxyphenyl)acetic acid
COC1=CC(=CC(=C1OC)OC)CC(=O)O
951-82-6
C11H14O5
226.23
MFCD00004336
DDSJXCGGOXKGSJ-UHFFFAOYSA-N
3,4,5-trimethoxyphenylacetic acid,2-3,4,5-trimethoxyphenyl acetic acid,3,4,5-trimethoxyphenyl acetic acid,benzeneacetic acid, 3,4,5-trimethoxy,3,4,5-trimethoxyphenylaceticacid,pubchem14015,acmc-209rv8,ksc487g4j,3,4,5-trimethoxybenzeneacetic acid,benzeneacetic acid,4,5-trimethoxy
70372
2-(3,4,5-trimethoxyphenyl)acetic acid
COC1=CC(=CC(=C1OC)OC)CC(=O)O
Quantity 10g
Packaging Glass bottle
Melting Point 117.0°C to 120.0°C
Infrared Spectrum Authentic
Color White to Cream
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3O)3C6H2CH2CO2H
Beilstein 10, 492
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone and in chloroform
Formula Weight 226.23
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 3, 4, 5-Trimethoxyphenylacetic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.