Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4,5-Trimethoxyphenylacetonitrile, 97%

Click to view available options
Quantity:
25g
13338-63-1
C11H13NO3
207.229
MFCD00001912
ACFJNTXCEQCDBX-UHFFFAOYSA-N
3,4,5-trimethoxyphenylacetonitrile,2-3,4,5-trimethoxyphenyl acetonitrile,3,4,5-trimethoxybenzyl cyanide,3,4,5-trimethoxybenzylnitrile,3,4,5-trimethoxybenzeneacetonitrile,benzeneacetonitrile, 3,4,5-trimethoxy,3,4,5-trimethoxyphenylacetylnitryl,3,4,5-trimethoxyphenyl acetonitrile,acetonitrile, 3,4,5-trimethoxyphenyl,2-3,4,5-trimethoxyphenyl ethanenitrile
25887
2-(3,4,5-trimethoxyphenyl)acetonitrile
COC1=CC(=CC(=C1OC)OC)CC#N
13338-63-1
C11H13NO3
207.229
MFCD00001912
ACFJNTXCEQCDBX-UHFFFAOYSA-N
3,4,5-trimethoxyphenylacetonitrile,2-3,4,5-trimethoxyphenyl acetonitrile,3,4,5-trimethoxybenzyl cyanide,3,4,5-trimethoxybenzylnitrile,3,4,5-trimethoxybenzeneacetonitrile,benzeneacetonitrile, 3,4,5-trimethoxy,3,4,5-trimethoxyphenylacetylnitryl,3,4,5-trimethoxyphenyl acetonitrile,acetonitrile, 3,4,5-trimethoxyphenyl,2-3,4,5-trimethoxyphenyl ethanenitrile
25887
2-(3,4,5-trimethoxyphenyl)acetonitrile
COC1=CC(=CC(=C1OC)OC)CC#N
Quantity 25g
Melting Point 75°C to 79°C
UN Number UN3439
Beilstein 2214548
Formula Weight 207.23
Percent Purity 97%
Chemical Name or Material 3,4,5-Trimethoxyphenylacetonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : AM2475000