Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,6-Tri-O-benzyl-D-galactal, ≥98%, Acros Organics

Click to view available options
Quantity:
1g
80040-79-5
C27H28O4
416.517
MFCD09039276
MXYLLYBWXIUMIT-ZONZVBGPSA-N
tri-o-benzyl-d-galactal,3,4,6-tri-o-benzyl-d-galactal,2r,3r,4r-3,4-bis benzyloxy-2-benzyloxy methyl-3,4-dihydro-2h-pyran,3-o,4-o,6-o-tribenzyl-1,2-dideoxy-d-lyxo-1-hexenopyranose,1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol 3,4,6-tribenzyl ether,2r,3r,4r-3,4-bis phenylmethoxy-2-phenylmethoxymethyl-3,4-dihydro-2h-pyran
2734741
(2R,3R,4R)-3,4-bis(phenylmethoxy)-2-(phenylmethoxymethyl)-3,4-dihydro-2H-pyran
C1=CC=C(C=C1)COCC2C(C(C=CO2)OCC3=CC=CC=C3)OCC4=CC=CC=C4
80040-79-5
C27H28O4
416.517
MFCD09039276
MXYLLYBWXIUMIT-ZONZVBGPSA-N
tri-o-benzyl-d-galactal,3,4,6-tri-o-benzyl-d-galactal,2r,3r,4r-3,4-bis benzyloxy-2-benzyloxy methyl-3,4-dihydro-2h-pyran,3-o,4-o,6-o-tribenzyl-1,2-dideoxy-d-lyxo-1-hexenopyranose,1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol 3,4,6-tribenzyl ether,2r,3r,4r-3,4-bis phenylmethoxy-2-phenylmethoxymethyl-3,4-dihydro-2h-pyran
2734741
(2R,3R,4R)-3,4-bis(phenylmethoxy)-2-(phenylmethoxymethyl)-3,4-dihydro-2H-pyran
C1=CC=C(C=C1)COCC2C(C(C=CO2)OCC3=CC=CC=C3)OCC4=CC=CC=C4
Quantity 1g
Color White to Yellow
Specific Rotation Condition −45° (20°C c=1,dichloromethane)
Infrared Spectrum Authentic
Specific Rotation -41° to -49° (20°C, 589nm) (c=1, dichloromethane)
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 416.52
Physical Form Crystalline Powder
Percent Purity ≥98.0%
Chemical Name or Material 3, 4, 6-Tri-O-benzyl-D-galactal
Show More Show Less