Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Bis(acetamido)benzoic acid, 97%

Click to view available options
Quantity:
1g
5g
205873-56-9
C11H12N2O4
236.227
MFCD00238577
GHTLCMPYQOFKCS-UHFFFAOYSA-N
3,4-bis acetylamino benzoic acid,benzoic acid,3,4-bis acetylamino,3,4-di acetylamino benzoic acid,maybridge1_006001,acmc-20anfw,3,4-bis acetamido benzoic acid
2735923
3,4-diacetamidobenzoic acid
CC(=O)NC1=C(C=C(C=C1)C(=O)O)NC(=O)C
205873-56-9
C11H12N2O4
236.227
MFCD00238577
GHTLCMPYQOFKCS-UHFFFAOYSA-N
3,4-bis acetylamino benzoic acid,benzoic acid,3,4-bis acetylamino,3,4-di acetylamino benzoic acid,maybridge1_006001,acmc-20anfw,3,4-bis acetamido benzoic acid
2735923
3,4-diacetamidobenzoic acid
CC(=O)NC1=C(C=C(C=C1)C(=O)O)NC(=O)C
Quantity 5g
Formula Weight 236.23
Percent Purity 97%
Chemical Name or Material 3,4-Bis(acetamido)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning