Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number AAH5193906
Click to view available options
Quantity:
1 g
5 g
883738-19-0
C22H35BN2O4
402.342
MFCD16294542
KBHWOVSSXKGKLY-UHFFFAOYSA-N
tert-butyl 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine-1-carboxylate, 3-4-boc-1-piperazinylmethyl benzeneboronic acid pinacol ester, tert-butyl 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine-1-carboxylate, amtb241, 3-4-boc-piperazine methyl phenylboronic acid pinacol ester, 3-4-boc-piperazine methyl phenylboronic-acid pinacol ester, 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl-1-piperazinecarboxylic acid tert-butyl ester
59298946
tert-butyl 4-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CN3CCN(CC3)C(=O)OC(C)(C)C
883738-19-0
C22H35BN2O4
402.342
MFCD16294542
KBHWOVSSXKGKLY-UHFFFAOYSA-N
tert-butyl 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine-1-carboxylate, 3-4-boc-1-piperazinylmethyl benzeneboronic acid pinacol ester, tert-butyl 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine-1-carboxylate, amtb241, 3-4-boc-piperazine methyl phenylboronic acid pinacol ester, 3-4-boc-piperazine methyl phenylboronic-acid pinacol ester, 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl-1-piperazinecarboxylic acid tert-butyl ester
59298946
tert-butyl 4-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CN3CCN(CC3)C(=O)OC(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Product of Advanced Molecular Technologies Pty Ltd.
TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 402.34
Percent Purity 95%
Chemical Name or Material 3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit