Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Bromobenzoyl)propionic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass Bottle
6340-79-0
C10H9BrO3
257.083
MFCD00016563
ZODFRCZNTXLDDW-UHFFFAOYSA-N
3-4-bromobenzoyl propionic acid,4-4-bromophenyl-4-oxobutanoic acid,3-p-bromobenzoyl propionic acid,4-4-bromo-phenyl-4-oxo-butyric acid,3-4-bromobenzoyl propanoic acid,4-4-bromophenyl-4-oxobutyric acid,4-4-bromophenyl-4-oxo-butyric acid,4-4-bromophenyl-4-oxo-butanoic acid,benzenebutanoic acid, 4-bromo-gamma-oxo,4-bromo-gamma-oxo-benzenebutanoic acid
80646
4-(4-bromophenyl)-4-oxobutanoic acid
C1=CC(=CC=C1C(=O)CCC(=O)O)Br
6340-79-0
C10H9BrO3
257.083
MFCD00016563
ZODFRCZNTXLDDW-UHFFFAOYSA-N
3-4-bromobenzoyl propionic acid,4-4-bromophenyl-4-oxobutanoic acid,3-p-bromobenzoyl propionic acid,4-4-bromo-phenyl-4-oxo-butyric acid,3-4-bromobenzoyl propanoic acid,4-4-bromophenyl-4-oxobutyric acid,4-4-bromophenyl-4-oxo-butyric acid,4-4-bromophenyl-4-oxo-butanoic acid,benzenebutanoic acid, 4-bromo-gamma-oxo,4-bromo-gamma-oxo-benzenebutanoic acid
80646
4-(4-bromophenyl)-4-oxobutanoic acid
C1=CC(=CC=C1C(=O)CCC(=O)O)Br
Quantity 25g
Packaging Glass Bottle
Color White to Beige
Melting Point 148.0°C to 150.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula BrC6H4CO(CH2)2COOH
Formula Weight 257.08
Physical Form Shiny Crystalline Flakes or Powder
Percent Purity 97%
Chemical Name or Material 3-(4-Bromobenzoyl)propionic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.