Alfa Aesar™ 3-(4-Bromophenyl)-1H-pyrazole, 97%

Manufacturer: Alfa Aesar™ H3322706

Quantity 5g

 View more versions of this product

Catalog No. AAH3322706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 73387-46-9
    Molecular Formula C9H7BrN2
    Formula Weight 223.07
    MDL Number MFCD01940434
    Chemical Name or Material 3-(4-Bromophenyl)-1H-pyrazole

    Specifications

    Formula Weight 223.07
    Molecular Formula C9H7BrN2
    MDL Number MFCD01940434
    CAS 73387-46-9
    Melting Point ≈130°C (polymorphic)
    Quantity 5g
    Chemical Name or Material 3-(4-Bromophenyl)-1H-pyrazole
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit