Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(4-Bromophenyl)-9-phenylcarbazole, 98%

Click to view available options
Quantity:
250mg
1g
5g
1028647-93-9
C24H16BrN
398.303
MFCD14582939
JEYLGFCAZBGCMC-UHFFFAOYSA-N
3-4-bromophenyl-9-phenyl-9h-carbazole,3-4-bromophenyl-9-phenylcarbazole,3-4-bromophenyl-n-phenylcarbazole,3-4-bromophenyl-9-phenyl9h-carbazole,pubchem23233,3-4-bromophenyl-9-phenyl-carbazole,3-4-bromo-phenyl-9-phenyl-9h-carbazole
51358293
3-(4-bromophenyl)-9-phenylcarbazole
C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52
1028647-93-9
C24H16BrN
398.303
MFCD14582939
JEYLGFCAZBGCMC-UHFFFAOYSA-N
3-4-bromophenyl-9-phenyl-9h-carbazole,3-4-bromophenyl-9-phenylcarbazole,3-4-bromophenyl-n-phenylcarbazole,3-4-bromophenyl-9-phenyl9h-carbazole,pubchem23233,3-4-bromophenyl-9-phenyl-carbazole,3-4-bromo-phenyl-9-phenyl-9h-carbazole
51358293
3-(4-bromophenyl)-9-phenylcarbazole
C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52
Quantity 250mg
Beilstein 19325695
Formula Weight 398.3
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-(4-Bromophenyl)-9-phenylcarbazole