Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(4-Chlorobenzoyl)propionic acid, 98%

Click to view available options
Quantity:
25g
100g
3984-34-7
C10H9ClO3
212.629
MFCD00002794
AHVASTJJVAYFPY-UHFFFAOYSA-N
4-4-chlorophenyl-4-oxobutanoic acid,3-4-chlorobenzoyl propionic acid,4-4-chloro-phenyl-4-oxo-butyric acid,3-4-chlorobenzoyl propanoic acid,3-p-chlorobenzoyl propionic acid,4-4-chlorophenyl-4-oxobutanoicacid,maybridge1_006214,ncistruc1_000312,ncistruc2_000239,asischem d51605
77604
4-(4-chlorophenyl)-4-oxobutanoic acid
C1=CC(=CC=C1C(=O)CCC(=O)O)Cl
3984-34-7
C10H9ClO3
212.629
MFCD00002794
AHVASTJJVAYFPY-UHFFFAOYSA-N
4-4-chlorophenyl-4-oxobutanoic acid,3-4-chlorobenzoyl propionic acid,4-4-chloro-phenyl-4-oxo-butyric acid,3-4-chlorobenzoyl propanoic acid,3-p-chlorobenzoyl propionic acid,4-4-chlorophenyl-4-oxobutanoicacid,maybridge1_006214,ncistruc1_000312,ncistruc2_000239,asischem d51605
77604
4-(4-chlorophenyl)-4-oxobutanoic acid
C1=CC(=CC=C1C(=O)CCC(=O)O)Cl
Melting Point 128°C to 130°C
Quantity 25g
Beilstein 2104409
Formula Weight 212.63
Percent Purity 98%
Chemical Name or Material 3-(4-Chlorobenzoyl)propionic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 223-627-3

TSCA : No

Recommended Storage : Ambient temperatures