Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Di-O-acetyl-L-arabinal, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
3945-18-4
C9H12O5
200.19
MFCD04112729
OWKCFBYWQGPLSJ-DTWKUNHWSA-N
3,4-di-o-acetyl-l-arabinal,3s,4s-3-acetyloxy-3,4-dihydro-2h-pyran-4-yl acetate,d-di-o-acetylarabinal,di-o-acetyl-d-arabinal,3,4-di-o-acetyl-d-ribal,3,4-dihydro-2h-pyran-3,4-diyl diacetate,1,2-dideoxy-d-erythro-pent-1-enopyranose diacetate,3s,4s-3,4-dihydro-2h-pyran-3,4-diyl diacetate,3s,4s-4-acetyloxy-3,4-dihydro-2h-pyran-3-yl acetate,1,5-anhydro-2-deoxy-d-erythro-pent-1-enitol 3,4-diacetate
8030588
[(3R,4S)-4-acetyloxy-3,4-dihydro-2H-pyran-3-yl] acetate
CC(=O)OC1COC=CC1OC(=O)C
3945-18-4
C9H12O5
200.19
MFCD04112729
OWKCFBYWQGPLSJ-DTWKUNHWSA-N
3,4-di-o-acetyl-l-arabinal,3s,4s-3-acetyloxy-3,4-dihydro-2h-pyran-4-yl acetate,d-di-o-acetylarabinal,di-o-acetyl-d-arabinal,3,4-di-o-acetyl-d-ribal,3,4-dihydro-2h-pyran-3,4-diyl diacetate,1,2-dideoxy-d-erythro-pent-1-enopyranose diacetate,3s,4s-3,4-dihydro-2h-pyran-3,4-diyl diacetate,3s,4s-4-acetyloxy-3,4-dihydro-2h-pyran-3-yl acetate,1,5-anhydro-2-deoxy-d-erythro-pent-1-enitol 3,4-diacetate
8030588
[(3R,4S)-4-acetyloxy-3,4-dihydro-2H-pyran-3-yl] acetate
CC(=O)OC1COC=CC1OC(=O)C
Refractive Index 1.4580 to 1.46
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Beilstein 17, IV, 2035
Formula Weight 200.19
Physical Form Oil
Percent Purity 97%
Chemical Name or Material 3, 4-Di-O-acetyl-L-arabinal
Show More Show Less