Alfa Aesar™ 3,4-Diacetamidobenzoic acid, 97%

Manufacturer: Alfa Aesar™ B2189003

Quantity 1g

 View more versions of this product

Catalog No. AAB2189003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 205873-56-9
    Molecular Formula C11H12N2O4
    Formula Weight 236.23
    MDL Number MFCD00238577
    Synonym 3,4-Bis(acetamido)benzoic acid
    Chemical Name or Material 3,4-Diacetamidobenzoic acid

    Specifications

    Formula Weight 236.23
    Melting Point 224° to 228°C
    Molecular Formula C11H12N2O4
    MDL Number MFCD00238577
    CAS 205873-56-9
    Synonym 3,4-Bis(acetamido)benzoic acid
    Quantity 1g
    Chemical Name or Material 3,4-Diacetamidobenzoic acid
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit