Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Diacetamidobenzoic acid, 97%

Click to view available options
Quantity:
1g
5g
205873-56-9
C11H12N2O4
236.227
MFCD00238577
GHTLCMPYQOFKCS-UHFFFAOYSA-N
3,4-bis acetylamino benzoic acid,benzoic acid,3,4-bis acetylamino,3,4-di acetylamino benzoic acid,maybridge1_006001,acmc-20anfw,3,4-bis acetamido benzoic acid
2735923
3,4-diacetamidobenzoic acid
CC(=O)NC1=C(C=C(C=C1)C(=O)O)NC(=O)C
205873-56-9
C11H12N2O4
236.227
MFCD00238577
GHTLCMPYQOFKCS-UHFFFAOYSA-N
3,4-bis acetylamino benzoic acid,benzoic acid,3,4-bis acetylamino,3,4-di acetylamino benzoic acid,maybridge1_006001,acmc-20anfw,3,4-bis acetamido benzoic acid
2735923
3,4-diacetamidobenzoic acid
CC(=O)NC1=C(C=C(C=C1)C(=O)O)NC(=O)C
Melting Point 224°C to 228°C
Quantity 5g
Formula Weight 236.23
Percent Purity 97%
Chemical Name or Material 3,4-Diacetamidobenzoic acid