Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Diacetoxybenzonitrile, 97%

Click to view available options
Quantity:
1g
5g
203626-45-3
C11H9NO4
219.196
OYRIDYHXQXSNCH-UHFFFAOYSA-N
3,4-diacetoxybenzonitrile,3,4-diacetoxy benzonitrile,4-cyano-1,2-phenylene diacetate,2-acetyloxy-5-cyanophenyl acetate
24721686
(2-acetyloxy-4-cyanophenyl) acetate
CC(=O)OC1=C(C=C(C=C1)C#N)OC(=O)C
203626-45-3
C11H9NO4
219.196
OYRIDYHXQXSNCH-UHFFFAOYSA-N
3,4-diacetoxybenzonitrile,3,4-diacetoxy benzonitrile,4-cyano-1,2-phenylene diacetate,2-acetyloxy-5-cyanophenyl acetate
24721686
(2-acetyloxy-4-cyanophenyl) acetate
CC(=O)OC1=C(C=C(C=C1)C#N)OC(=O)C
Melting Point 87°C
Quantity 1g
Color White
UN Number UN3439
Formula Weight 219.19
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 3,4-Diacetoxybenzonitrile

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.