Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Diamino-5-bromopyridine, 96%

Click to view available options
Quantity:
250mg
1g
4635-08-9
C5H6BrN3
188.03
MFCD01692479
CMFSWSZEHYPECE-UHFFFAOYSA-N
5-bromo-3,4-diaminopyridine,pyridine, 3-bromo-4,5-diamino,5-bromo-pyridine-3,4-diamine,3-bromo-4,5-diaminopyridine,3,4-pyridinediamine, 5-bromo,5-bromo-3,4-pyridinediamine,pubchem23608,3,4-diamino-5-bromopyridine,3-bromo-4,5-pyridinediamine,3,4-diamino-5-bromopyridine
78369
5-bromopyridine-3,4-diamine
C1=C(C(=C(C=N1)Br)N)N
4635-08-9
C5H6BrN3
188.03
MFCD01692479
CMFSWSZEHYPECE-UHFFFAOYSA-N
5-bromo-3,4-diaminopyridine,pyridine, 3-bromo-4,5-diamino,5-bromo-pyridine-3,4-diamine,3-bromo-4,5-diaminopyridine,3,4-pyridinediamine, 5-bromo,5-bromo-3,4-pyridinediamine,pubchem23608,3,4-diamino-5-bromopyridine,3-bromo-4,5-pyridinediamine,3,4-diamino-5-bromopyridine
78369
5-bromopyridine-3,4-diamine
C1=C(C(=C(C=N1)Br)N)N
Melting Point 138°C to 145°C
Color White to Yellow
Quantity 250mg
Sensitivity Moisture & Light Sensitive
UN Number UN2811
Formula Weight 188.03
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 3,4-Diamino-5-bromopyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : US4000000