Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Diamino-5-bromopyridine, 96%

Click to view available options
Quantity:
250mg
1g
4653-11-6
C8H9O2S
169.22
MFCD01692479
VYTXLSQVYGNWLV-UHFFFAOYSA-M
4-2-thienyl butyric acid,4-thiophen-2-yl butanoic acid,2-thiophenebutanoic acid,2-thiophenebutyric acid,4-2-thienyl butanoic acid,4-thiophen-2-yl-butyric acid,4-thiophen-2-yl-butyricacid,gamma-alpha-thienyl butyricacid,.gamma.-.alpha.-thienyl butyric acid,4tb
78386
CHEBI:40114
4-thiophen-2-ylbutanoic acid
C1=CSC(=C1)CCCC(=O)O
4653-11-6
C8H9O2S
169.22
MFCD01692479
VYTXLSQVYGNWLV-UHFFFAOYSA-M
4-2-thienyl butyric acid,4-thiophen-2-yl butanoic acid,2-thiophenebutanoic acid,2-thiophenebutyric acid,4-2-thienyl butanoic acid,4-thiophen-2-yl-butyric acid,4-thiophen-2-yl-butyricacid,gamma-alpha-thienyl butyricacid,.gamma.-.alpha.-thienyl butyric acid,4tb
78386
CHEBI:40114
4-thiophen-2-ylbutanoic acid
C1=CSC(=C1)CCCC(=O)O
Color White to Yellow
Sensitivity Moisture & Light Sensitive
Quantity 1g
Melting Point 138°C to 145°C
UN Number UN2811
Formula Weight 188.03
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 3,4-Diamino-5-bromopyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : US4000000