Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Diaminobenzamidoxime, 97%

Click to view available options
Quantity:
250mg
1g
1469816-70-3
C7H10N4O
166.184
OPOKBHCWRPVYFH-UHFFFAOYSA-N
3,4-diaminobenzamidoxime,3,4-diamino-n-hydroxybenzimidamide
86277480
3,4-diamino-N'-hydroxybenzenecarboximidamide
C1=CC(=C(C=C1C(=NO)N)N)N
1469816-70-3
C7H10N4O
166.184
OPOKBHCWRPVYFH-UHFFFAOYSA-N
3,4-diaminobenzamidoxime,3,4-diamino-n-hydroxybenzimidamide
86277480
3,4-diamino-N'-hydroxybenzenecarboximidamide
C1=CC(=C(C=C1C(=NO)N)N)N
Quantity 1g
Color Brown
Formula Weight 166.18
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3,4-Diaminobenzamidoxime