Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Diaminobenzonitrile, 97%

Click to view available options
Quantity:
1g
5g
25g
17626-40-3
C7H7N3
133.154
MFCD00723901
VWLLPPSBBHDXHK-UHFFFAOYSA-N
3,4-diamino-benzonitrile,benzonitrile, 3,4-diamino,4-cyanobenzene-1,2-diamine,3,4-diaminobenzenecarbonitrile,zlchem 619,pubchem2547,3, 4-diaminobenzonitrile,3,4 diamino benzonitrile,3,4-diamino benzonitrile,acmc-209eb5
737285
3,4-diaminobenzonitrile
C1=CC(=C(C=C1C#N)N)N
17626-40-3
C7H7N3
133.154
MFCD00723901
VWLLPPSBBHDXHK-UHFFFAOYSA-N
3,4-diamino-benzonitrile,benzonitrile, 3,4-diamino,4-cyanobenzene-1,2-diamine,3,4-diaminobenzenecarbonitrile,zlchem 619,pubchem2547,3, 4-diaminobenzonitrile,3,4 diamino benzonitrile,3,4-diamino benzonitrile,acmc-209eb5
737285
3,4-diaminobenzonitrile
C1=CC(=C(C=C1C#N)N)N
Applications
3,4-Diaminobenzonitrile is usually used like synthetic intermediate.

Solubility
Soluble in dichloromethane, methanol and ethyl acetate.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Color Brown
Quantity 25g
Melting Point 140°C to 148°C
UN Number UN3439
Formula Weight 133.15
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3,4-Diaminobenzonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.