Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diaminobenzophenone, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
39070-63-8
C13H12N2O
212.252
MFCD00007727
RXCOGDYOZQGGMK-UHFFFAOYSA-N
3,4-diaminobenzophenone,3,4-diaminophenyl phenyl methanone,3,4-diaminophenyl phenylmethanone,4-benzoylbenzene-1,2-diamine,2-amino-4-benzoylaniline,4-benzoyl-o-phenylenediamine,3,4-diaminophenyl phenyl ketone,3,4-diaminophenyl-phenylmethanone,methanone, 3,4-diaminophenyl phenyl,pubchem3382
135520
(3,4-diaminophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)N)N
39070-63-8
C13H12N2O
212.252
MFCD00007727
RXCOGDYOZQGGMK-UHFFFAOYSA-N
3,4-diaminobenzophenone,3,4-diaminophenyl phenyl methanone,3,4-diaminophenyl phenylmethanone,4-benzoylbenzene-1,2-diamine,2-amino-4-benzoylaniline,4-benzoyl-o-phenylenediamine,3,4-diaminophenyl phenyl ketone,3,4-diaminophenyl-phenylmethanone,methanone, 3,4-diaminophenyl phenyl,pubchem3382
135520
(3,4-diaminophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)N)N
Color Ochre-Yellow to Yellow
Sulfated Ash 0.3% max.
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Quantity 100g
Melting Point 116°C to 119°C
Packaging Plastic bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5COC6H3(NH2)2
Beilstein 14, II, 225
Formula Weight 212.25
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 3, 4-Diaminobenzophenone, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 254-273-8

TSCA : TSCA