Alfa Aesar™ 3,4-Diaminotoluene, 97%

Manufacturer: Alfa Aesar™ B2437822

Quantity 100g

 View more versions of this product

Catalog No. AAB2437822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 496-72-0
    Molecular Formula C7H10N2
    Formula Weight 122.17
    MDL Number MFCD00007728
    Beilstein 507965
    Synonym 4-Methyl-1,2-phenylenediamine; 3,4-Toluenediamine
    Chemical Name or Material 3,4-Diaminotoluene

    Specifications

    Formula Weight 122.17
    MDL Number MFCD00007728
    Health Hazard 2 GHS H Statement
    H301-H341-H319
    Toxic if swallowed.
    Suspected of causing genetic defects.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P281-P301+P310-P305+P351+P338-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 265°C
    Melting Point 86° to 90°C
    Quantity 100g
    TSCA TSCA
    Molecular Formula C7H10N2
    CAS 496-72-0
    Beilstein 507965
    Synonym 4-Methyl-1,2-phenylenediamine; 3,4-Toluenediamine
    Health Hazard 1 Danger
    RTECS Number XS9820000
    Chemical Name or Material 3,4-Diaminotoluene
    Assay 97%

    GHS H Statement
    H301-H341-H319
    Toxic if swallowed.
    Suspected of causing genetic defects.
    Causes serious eye irritation.

    GHS P Statement
    P280-P281-P301+P310-P305+P351+P338-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit