Alfa Aesar™ 3,4-Dichloro-1,2,5-thiadiazole, 99%

Manufacturer: Alfa Aesar™ A1173818

Quantity 50g

 View more versions of this product

Catalog No. AAA1173818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5728-20-1
    Molecular Formula C2Cl2N2S
    Formula Weight 155.01
    MDL Number MFCD00010072
    UN Number UN2810
    Beilstein 606585
    Chemical Name or Material 3,4-Dichloro-1,2,5-thiadiazole

    Specifications

    Formula Weight 155.01
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00010072
    CAS 5728-20-1
    Beilstein 606585
    Flash Point >100°C (212°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Molecular Formula C2Cl2N2S
    UN Number UN2810
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Quantity 50g
    Boiling Point 157° to 159°C
    Chemical Name or Material 3,4-Dichloro-1,2,5-thiadiazole
    Assay 99%

    Description

    Applications
    3,4-Dichloro-1,2,5-thiadiazole is a chlorinated thiadiazole that is a potent inhibitors of nitrification in soil. 3,4-Dichloro-1,2,5-thiadiazole is used as a building block in the preparaton of the β-adrenergic blocking agent, Timolol.

    Solubility
    Soluble in chloroform and methanol.

    Notes
    Hygroscopic. Store under dry inert gas. Incompatible with moisture and oxidizing agents, acids and bases.

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit