Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzoic acid, 98+%, ACROS Organics™

Click to view available options
Quantity:
100g
Packaging:
Plastic bottle
51-44-5
C7H4Cl2O2
191.007
MFCD00002492
VPHHJAOJUJHJKD-UHFFFAOYSA-N
benzoic acid, 3,4-dichloro,unii-ys4zr9bxwx,3,4-dichloro-benzoic acid,ys4zr9bxwx,benzoic acid,4-dichloro,wln: qvr cg dg,pubchem3686,acmc-209kts,dsstox_cid_4980,dsstox_rid_77609
5817
CHEBI:49401
3,4-dichlorobenzoic acid
C1=CC(=C(C=C1C(=O)O)Cl)Cl
51-44-5
C7H4Cl2O2
191.007
MFCD00002492
VPHHJAOJUJHJKD-UHFFFAOYSA-N
benzoic acid, 3,4-dichloro,unii-ys4zr9bxwx,3,4-dichloro-benzoic acid,ys4zr9bxwx,benzoic acid,4-dichloro,wln: qvr cg dg,pubchem3686,acmc-209kts,dsstox_cid_4980,dsstox_rid_77609
5817
CHEBI:49401
3,4-dichlorobenzoic acid
C1=CC(=C(C=C1C(=O)O)Cl)Cl
Infrared Spectrum Authentic
Quantity 100g
Packaging Plastic bottle
Melting Point 205°C to 209°C
Color White to Beige
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula Cl2C6H3CO2H
Formula Weight 191.01
Physical Form Powder
Percent Purity ≥98.0%
Chemical Name or Material 3, 4-Dichlorobenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.