Alfa Aesar™ 3,4-Dichlorobenzophenone, 99%

Manufacturer: Alfa Aesar™ B2294736

Quantity 500g

 View more versions of this product

Catalog No. AAB2294736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6284-79-3
    Molecular Formula C13H8Cl2O
    Formula Weight 251.11
    MDL Number MFCD00000552
    Beilstein 2105069
    Chemical Name or Material 3,4-Dichlorobenzophenone

    Specifications

    Formula Weight 251.11
    Odor Odorless
    Melting Point 100° to 104°C
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 500g
    Molecular Formula C13H8Cl2O
    MDL Number MFCD00000552
    CAS 6284-79-3
    Beilstein 2105069
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 3,4-Dichlorobenzophenone
    Assay 99%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit