Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dichlorobenzoyl chloride, 97%

Click to view available options
Quantity:
100g
3024-72-4
C7H3Cl3O
209.45
MFCD00000672
VTXNOVCTHUBABW-UHFFFAOYSA-N
benzoyl chloride, 3,4-dichloro,3,4-dichlorobenzoylchloride,3,4-dichloro benzoyl chloride,2,3-dichlorobenzoyl,3,4dichlorobenzoyl chloride,acmc-1aj45,3,4 dichlorobenzoyl chloride,3,4-dichlorobenzoyl-chloride,ksc225o5d,3,4-dichloro-benzoyl chloride
76403
3,4-dichlorobenzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)Cl)Cl
3024-72-4
C7H3Cl3O
209.45
MFCD00000672
VTXNOVCTHUBABW-UHFFFAOYSA-N
benzoyl chloride, 3,4-dichloro,3,4-dichlorobenzoylchloride,3,4-dichloro benzoyl chloride,2,3-dichlorobenzoyl,3,4dichlorobenzoyl chloride,acmc-1aj45,3,4 dichlorobenzoyl chloride,3,4-dichlorobenzoyl-chloride,ksc225o5d,3,4-dichloro-benzoyl chloride
76403
3,4-dichlorobenzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)Cl)Cl
Sensitivity Moisture sensitive
Boiling Point 242°C
Melting Point 30°C to 34°C
Flash Point 142°C (287°F)
Quantity 100g
UN Number UN3261
Beilstein 607485
Formula Weight 209.46
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzoyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 221-177-2

TSCA : Yes

Recommended Storage : Ambient temperatures