Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzoyl Chloride 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3024-72-4
C7H3Cl3O
209.45
MFCD00000672
VTXNOVCTHUBABW-UHFFFAOYSA-N
benzoyl chloride, 3,4-dichloro,3,4-dichlorobenzoylchloride,3,4-dichloro benzoyl chloride,2,3-dichlorobenzoyl,3,4dichlorobenzoyl chloride,acmc-1aj45,3,4 dichlorobenzoyl chloride,3,4-dichlorobenzoyl-chloride,ksc225o5d,3,4-dichloro-benzoyl chloride
76403
3,4-dichlorobenzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)Cl)Cl
3024-72-4
C7H3Cl3O
209.45
MFCD00000672
VTXNOVCTHUBABW-UHFFFAOYSA-N
benzoyl chloride, 3,4-dichloro,3,4-dichlorobenzoylchloride,3,4-dichloro benzoyl chloride,2,3-dichlorobenzoyl,3,4dichlorobenzoyl chloride,acmc-1aj45,3,4 dichlorobenzoyl chloride,3,4-dichlorobenzoyl-chloride,ksc225o5d,3,4-dichloro-benzoyl chloride
76403
3,4-dichlorobenzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)Cl)Cl
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula Cl2C6H3COCl
Beilstein 09, 344
Formula Weight 209.46
Physical Form Crystalline Low Melting Solid
Percent Purity ≥96.0% (RCOCl)
Chemical Name or Material 3, 4-Dichlorobenzoyl chloride, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-177-2

TSCA : TSCA