Alfa Aesar™ 3,4-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF

Manufacturer: Alfa Aesar™ H54133AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH54133AE

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 61259-69-6
    Molecular Formula C7H5Cl3Mg
    Formula Weight 219.78
    MDL Number MFCD01311472
    UN Number UN2924
    Chemical Name or Material 3,4-Dichlorobenzylmagnesium chloride

    Specifications

    Formula Weight 219.78
    Molecular Formula C7H5Cl3Mg
    MDL Number MFCD01311472
    CAS 61259-69-6
    UN Number UN2924
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Concentration 0.25M in 2-MeTHF
    Quantity 100mL
    Chemical Name or Material 3,4-Dichlorobenzylmagnesium chloride

    Description

    Applications
    3,4-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF is used as a pharmaceutical intermediate.

    Solubility
    Reacts with water.

    Notes
    Air and moisture sensitive. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.

    GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit