Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dichlorocinnamic acid, 97%

Click to view available options
Quantity:
5g
25g
100g
1202-39-7
C9H6Cl2O2
217.045
MFCD00004385
RRLUFPHCTSFKNR-DUXPYHPUSA-N
3,4-dichlorocinnamic acid,e-3-3,4-dichlorophenyl acrylic acid,3',4'-dichlorocinnamic acid,2-propenoic acid, 3-3,4-dichlorophenyl,cinnamic acid, 3,4-dichloro,3-3,4-dichlorophenyl acrylic acid,2e-3-3,4-dichlorophenyl prop-2-enoic acid,3-3,4-dichlorophenyl-2-propenoic acid,e-3-3,4-dichlorophenyl prop-2-enoic acid,2e-3-3,4-dichlorophenyl acrylic acid
688027
(E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1C=CC(=O)O)Cl)Cl
1202-39-7
C9H6Cl2O2
217.045
MFCD00004385
RRLUFPHCTSFKNR-DUXPYHPUSA-N
3,4-dichlorocinnamic acid,e-3-3,4-dichlorophenyl acrylic acid,3',4'-dichlorocinnamic acid,2-propenoic acid, 3-3,4-dichlorophenyl,cinnamic acid, 3,4-dichloro,3-3,4-dichlorophenyl acrylic acid,2e-3-3,4-dichlorophenyl prop-2-enoic acid,3-3,4-dichlorophenyl-2-propenoic acid,e-3-3,4-dichlorophenyl prop-2-enoic acid,2e-3-3,4-dichlorophenyl acrylic acid
688027
(E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1C=CC(=O)O)Cl)Cl
Melting Point 218°C to 220°C
Quantity 100g
Beilstein 1872129
Formula Weight 217.05
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorocinnamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UD3333900