Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorophenethylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
21581-45-3
C8H9Cl2N
190.07
MFCD00060617
MQPUAVYKVIHUJP-UHFFFAOYSA-N
3,4-dichlorophenethylamine,2-3,4-dichlorophenyl ethanamine,2-3,4-dichlorophenyl ethylamine,2-3,4-dichlorophenyl ethan-1-amine,2-3,4-dichloro-phenyl-ethylamine,benzeneethanamine, 3,4-dichloro,2-3,4,-dichlorophenyl ethylamine,1-amino-2-3,4-dichlorophenyl ethane,pubchem12521,acmc-1apis
217958
2-(3,4-dichlorophenyl)ethanamine
C1=CC(=C(C=C1CCN)Cl)Cl
21581-45-3
C8H9Cl2N
190.07
MFCD00060617
MQPUAVYKVIHUJP-UHFFFAOYSA-N
3,4-dichlorophenethylamine,2-3,4-dichlorophenyl ethanamine,2-3,4-dichlorophenyl ethylamine,2-3,4-dichlorophenyl ethan-1-amine,2-3,4-dichloro-phenyl-ethylamine,benzeneethanamine, 3,4-dichloro,2-3,4,-dichlorophenyl ethylamine,1-amino-2-3,4-dichlorophenyl ethane,pubchem12521,acmc-1apis
217958
2-(3,4-dichlorophenyl)ethanamine
C1=CC(=C(C=C1CCN)Cl)Cl
Specific Gravity 1.268
Density 1.2680g/mL
Refractive Index 1.5655 to 1.5675
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Beilstein 12, IV, 2491
Formula Weight 190.07
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3, 4-Dichlorophenethylamine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger