Alfa Aesar™ 3,4-Dichlorophenol, 99%

Manufacturer: Alfa Aesar™ A1225614

Quantity 25g

 View more versions of this product

Catalog No. AAA1225614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 95-77-2
    Molecular Formula C6H4Cl2O
    Formula Weight 163
    MDL Number MFCD00002258
    UN Number UN2020
    Beilstein 1907693
    Chemical Name or Material 3,4-Dichlorophenol

    Specifications

    Formula Weight 163
    Health Hazard 3 GHS P Statement
    P280h-P273-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Molecular Formula C6H4Cl2O
    CAS 95-77-2
    Health Hazard 1 Warning
    Boiling Point 252° to 255°C
    Beilstein 1907693
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
    RTECS Number SK8800000
    Quantity 25g
    TSCA TSCA
    Odor Phenol-like
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 64° to 68°C
    MDL Number MFCD00002258
    UN Number UN2020
    Chemical Name or Material 3,4-Dichlorophenol
    Assay 99%

    Description

    Applications
    It is also a photo-degradation product of the common antibacterial and antifungal agent triclosan along with the dioxin 2,8-dichlorodibenzo-p-dioxin.

    Solubility
    Slightly soluble ion water 9.2 g/L at 20degC. Soluble in aqueous alkali, oxygenated and chlorinated solvents. Soluble in ethanol, benzene, chloroform and ethyl ether.

    Notes
    Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, acid, acid fumes, acid chlorides, acid anhydrides, caustics. Keep container tightly closed. Store in an area without drain or sewer access. Separated from strong oxidants and food and feedstuffs.

    GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P273-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit