Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dichlorophenylacetic acid, 98%

Click to view available options
Quantity:
5g
25g
100g
5807-30-7
C8H6Cl2O2
205.034
MFCD00004333
ZOUPGSMSNQLUNW-UHFFFAOYSA-N
3,4-dichlorophenylacetic acid,2-3,4-dichlorophenyl acetic acid,3,4-dichlorophenyl acetic acid,3,4-dichlorobenzene acetic acid,benzeneacetic acid, 3,4-dichloro,pubchem20195,acmc-209m2g,3,4dichlorophenylacetic acid,3,4-dichlorophenylaceticacid
79874
2-(3,4-dichlorophenyl)acetic acid
C1=CC(=C(C=C1CC(=O)O)Cl)Cl
5807-30-7
C8H6Cl2O2
205.034
MFCD00004333
ZOUPGSMSNQLUNW-UHFFFAOYSA-N
3,4-dichlorophenylacetic acid,2-3,4-dichlorophenyl acetic acid,3,4-dichlorophenyl acetic acid,3,4-dichlorobenzene acetic acid,benzeneacetic acid, 3,4-dichloro,pubchem20195,acmc-209m2g,3,4dichlorophenylacetic acid,3,4-dichlorophenylaceticacid
79874
2-(3,4-dichlorophenyl)acetic acid
C1=CC(=C(C=C1CC(=O)O)Cl)Cl
Applications
It is used in the parallel solid phase synthesis of tetra substituted diethylenetriamines via selective amide alkylation and exhaustive reduction of N-acylated dipeptides.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point 87°C to 92°C
Odor Odorless
Quantity 25g
Beilstein 1953781
Solubility Information Insoluble in water.
Formula Weight 205.04
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorophenylacetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 227-368-7

TSCA : No

Recommended Storage : Ambient temperatures