Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorophenylacetonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
3218-49-3
C8H5Cl2N
186.04
MFCD00001909
QWZNCAFWRZZJMA-UHFFFAOYSA-N
3,4-dichlorophenylacetonitrile,2-3,4-dichlorophenyl acetonitrile,3,4-dichlorobenzyl cyanide,3,4-dichlorophenyl acetonitrile,benzeneacetonitrile, 3,4-dichloro,3,4-dichlorobenzeneacetonitrile,2-3,4-dichlorophenyl ethanenitrile,3,4-dichlorphenyl acetonitril,pubchem16506,3,4-dichlorobenzylcyanide
76690
2-(3,4-dichlorophenyl)acetonitrile
C1=CC(=C(C=C1CC#N)Cl)Cl
3218-49-3
C8H5Cl2N
186.04
MFCD00001909
QWZNCAFWRZZJMA-UHFFFAOYSA-N
3,4-dichlorophenylacetonitrile,2-3,4-dichlorophenyl acetonitrile,3,4-dichlorobenzyl cyanide,3,4-dichlorophenyl acetonitrile,benzeneacetonitrile, 3,4-dichloro,3,4-dichlorobenzeneacetonitrile,2-3,4-dichlorophenyl ethanenitrile,3,4-dichlorphenyl acetonitril,pubchem16506,3,4-dichlorobenzylcyanide
76690
2-(3,4-dichlorophenyl)acetonitrile
C1=CC(=C(C=C1CC#N)Cl)Cl
Flash Point >110°C
Packaging Glass bottle
Melting Point 38.0°C to 41.0°C
Boiling Point 170.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Color White
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula Cl2C6H3CH2CN
Beilstein 09, III, 2272
Formula Weight 186.04
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorophenylacetonitrile, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-743-9

TSCA : TSCA