Learn More
Alfa Aesar™ 3,4-Dichlorophenylmagnesium bromide, 0.50M in 2-MeTHF
Manufacturer:  Alfa Aesar™ H54621AE
Quantity 100mL
 View more versions of this product
Catalog No. AAH54621AE
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
79175-35-2
250.2
UN2924
C6H3BrCl2Mg
MFCD01319891
3,4-Dichlorophenylmagnesium bromide
Specifications
0.50M in 2-MeTHF
79175-35-2
250.2
UN2924
100mL
C6H3BrCl2Mg
MFCD01319891
3,4-Dichlorophenylmagnesium bromide
Safety and Handling
GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.