3,4-Difluoro-2-methylbenzoic acid, 97+%, Alfa Aesar™

Manufacturer: Alfa Aesar H3232603

Quantity 1g

 View more versions of this product

Catalog No. AAH3232603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 157652-31-8
    Molecular Formula C8H6F2O2
    Formula Weight 172.13
    MDL Number MFCD07777153
    Synonym 3,4-Difluoro-o-toluic acid
    Chemical Name or Material 3,4-Difluoro-2-methylbenzoic acid

    Specifications

    Formula Weight 172.13
    Quantity 1g
    MDL Number MFCD07777153
    CAS 157652-31-8
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color White to Cream
    Melting Point 152° to 156°C
    Physical Form Powder
    Molecular Formula C8H6F2O2
    Synonym 3,4-Difluoro-o-toluic acid
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Odor Odorless
    Chemical Name or Material 3,4-Difluoro-2-methylbenzoic acid
    Assay ≥97%

    Description

    Applications
    3,4-Difluoro-2-methylbenzoic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in medicine field, electronic industry and polymer material.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit