Alfa Aesar™ 3,4-Difluoro-2-methylbenzonitrile, 98%

Manufacturer: Alfa Aesar™ H3154806

Quantity 5g

 View more versions of this product

Catalog No. AAH3154806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 847502-83-4
    Molecular Formula C8H5F2N
    Formula Weight 153.13
    MDL Number MFCD07777154
    UN Number UN3439
    Synonym 3,4-Difluoro-o-tolunitrile
    Chemical Name or Material 3,4-Difluoro-2-methylbenzonitrile

    Specifications

    Formula Weight 153.13
    Molecular Formula C8H5F2N
    MDL Number MFCD07777154
    CAS 847502-83-4
    UN Number UN3439
    Synonym 3,4-Difluoro-o-tolunitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Melting Point 39° to 44°C
    Quantity 5g
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 3,4-Difluoro-2-methylbenzonitrile
    Assay 98%

    GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit