Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Difluorobenzyl alcohol, 99%

Click to view available options
Quantity:
1g
5g
85118-05-4
C7H6F2O
144.121
MFCD00010628
GNQLTCVBSGVGHC-UHFFFAOYSA-N
3,4-difluorobenzyl alcohol,3,4-difluorophenyl methanol,benzenemethanol, 3,4-difluoro,3,4-difluorobenzylalcohol,3,4-difluoro-phenyl-methanol,3,4-difluorophenyl methan-1-ol,3, 4-difluorobenzyl alcohol,pubchem4916,acmc-209q4o,ksc493q4f
522833
(3,4-difluorophenyl)methanol
C1=CC(=C(C=C1CO)F)F
85118-05-4
C7H6F2O
144.121
MFCD00010628
GNQLTCVBSGVGHC-UHFFFAOYSA-N
3,4-difluorobenzyl alcohol,3,4-difluorophenyl methanol,benzenemethanol, 3,4-difluoro,3,4-difluorobenzylalcohol,3,4-difluoro-phenyl-methanol,3,4-difluorophenyl methan-1-ol,3, 4-difluorobenzyl alcohol,pubchem4916,acmc-209q4o,ksc493q4f
522833
(3,4-difluorophenyl)methanol
C1=CC(=C(C=C1CO)F)F
Flash Point 97°C (206°F)
Quantity 1g
Refractive Index 1.49
Density 1.262
Beilstein 9043426
Formula Weight 144.12
Percent Purity 99%
Chemical Name or Material 3,4-Difluorobenzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 285-657-3

TSCA : No

Recommended Storage : Ambient temperatures