Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluorophenyl isocyanate, ACROS Organics™

Click to view available options
Quantity:
500mg
10g
Packaging:
Glass Bottle in Plastic Container
42601-04-7
C7H3F2NO
155.104
MFCD00065711
YBWJVFMNWMLZEN-UHFFFAOYSA-N
3,4-difluorophenyl isocyanate,3,4-difluorophenylisocyanate,isocyanic acid 3,4-difluorophenyl ester,3,4-difluoro phenyl isocyanate,benzene,1,2-difluoro-4-isocyanato,1,2-difluoro-4-isocyanato-benzene,3,4-difluorobenzenisocyanate,acmc-1apvi,3,4-difluoro-phenylisocyanate,3,4-difluoro-phenyl isocyanate
2734708
1,2-difluoro-4-isocyanatobenzene
C1=CC(=C(C=C1N=C=O)F)F
42601-04-7
C7H3F2NO
155.104
MFCD00065711
YBWJVFMNWMLZEN-UHFFFAOYSA-N
3,4-difluorophenyl isocyanate,3,4-difluorophenylisocyanate,isocyanic acid 3,4-difluorophenyl ester,3,4-difluoro phenyl isocyanate,benzene,1,2-difluoro-4-isocyanato,1,2-difluoro-4-isocyanato-benzene,3,4-difluorobenzenisocyanate,acmc-1apvi,3,4-difluoro-phenylisocyanate,3,4-difluoro-phenyl isocyanate
2734708
1,2-difluoro-4-isocyanatobenzene
C1=CC(=C(C=C1N=C=O)F)F
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Quantity 10g
Packaging Glass Bottle in Plastic Container
Color Yellow
Infrared Spectrum Authentic
Density 1.3260g/mL
Linear Formula F2C6H3NCO
Formula Weight 155.1
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3, 4-Difluorophenyl isocyanate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C