Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Difluorophenylacetic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
658-93-5
C8H6F2O2
172.131
MFCD00010002
YCAKYFIYUHHCKW-UHFFFAOYSA-N
3,4-difluorophenylacetic acid,2-3,4-difluorophenyl acetic acid,3,4-difluorophenyl acetic acid,benzeneacetic acid, 3,4-difluoro,3,4-difluorobenzeneacetic acid,3,4-difluorophenylaceticacid,pubchem4154,acmc-209nst,ksc355a2l,rarechem al bo 0250
69571
2-(3,4-difluorophenyl)acetic acid
C1=CC(=C(C=C1CC(=O)O)F)F
658-93-5
C8H6F2O2
172.131
MFCD00010002
YCAKYFIYUHHCKW-UHFFFAOYSA-N
3,4-difluorophenylacetic acid,2-3,4-difluorophenyl acetic acid,3,4-difluorophenyl acetic acid,benzeneacetic acid, 3,4-difluoro,3,4-difluorobenzeneacetic acid,3,4-difluorophenylaceticacid,pubchem4154,acmc-209nst,ksc355a2l,rarechem al bo 0250
69571
2-(3,4-difluorophenyl)acetic acid
C1=CC(=C(C=C1CC(=O)O)F)F
Melting Point 48°C to 50°C
Quantity 1g
Flash Point >110°C (230°F)
Beilstein 2577768
Formula Weight 172.13
Percent Purity 98%
Chemical Name or Material 3,4-Difluorophenylacetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-527-2

TSCA : No

Recommended Storage : Ambient temperatures