Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Description
3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Reacts with water.
3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Reacts with water.
Specifications
Quantity | 100 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive; May form a precipitate |
Solubility Information | Reacts with water. |
Formula Weight | 217.29 |
Concentration or Composition (by Analyte or Components) | 0.50M in 2-MeTHF |
Chemical Name or Material | 3,4-Difluorophenylmagnesium bromide |
Safety and Handling
Hazard Category | H225-H302-H314-H335 |
Hazard Statement | GHS H Statement H225-H314 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction