Alfa Aesar™ 3,4-Dihydro-1H-isoquinoline-2-carboxamidine hydriodide, 98%

Manufacturer: Alfa Aesar™ H2773103

Quantity 1g

 View more versions of this product

Catalog No. AAH2773103

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C10H13N3·HI
    Formula Weight 303.14
    MDL Number MFCD07681953
    Synonym Debrisoquin hydriodide
    Chemical Name or Material 3,4-Dihydro-1H-isoquinoline-2-carboxamidine hydriodide

    Specifications

    Formula Weight 303.14
    Molecular Formula C10H13N3·HI
    MDL Number MFCD07681953
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    Melting Point 152° to 154°C
    Synonym Debrisoquin hydriodide
    Chemical Name or Material 3,4-Dihydro-1H-isoquinoline-2-carboxamidine hydriodide
    Assay 98%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit