Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dihydro-2-(1H)-quinolinone, 98%

Manufacturer:  Alfa Aesar™ H3663814

Quantity 25g

 View more versions of this product

Catalog No. AAH3663814


  Check Availability
Add to cart

Specifications

Description

Applications
3,4-Dihydro-2-(1H)-quinolinone may be employed as medium supplement in the culture medium of Pseudomonas ayucida during enrichment culture experiments. It is used to prepare potent bicyclic peptide deformylase inhibitors with antibacterial effects. It is also used to synthesize substituted iminopiperidines as inhibitors of human nitric oxide synthase isoforms.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

553-03-7
147.18
Hydrocarbostyril
C9H9NO
MFCD00016722
3,4-Dihydro-2-(1H)-quinolinone

Specifications

25g
C9H9NO
MFCD00016722
Hydrocarbostyril
98%
553-03-7
147.18
165° to 167°C
3,4-Dihydro-2-(1H)-quinolinone

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit