Alfa Aesar™ 3,4-Dihydro-2-methoxy-2H-pyran, 99%

Manufacturer: Alfa Aesar™ B2460922

Quantity 100g

 View more versions of this product

Catalog No. AAB2460922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4454-05-1
    Molecular Formula C6H10O2
    Formula Weight 114.14
    MDL Number MFCD00006560
    UN Number UN3271
    Beilstein 107274
    Chemical Name or Material 3,4-Dihydro-2-methoxy-2H-pyran

    Specifications

    Formula Weight 114.14
    TSCA TSCA
    MDL Number MFCD00006560
    UN Number UN3271
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H301-H315-H319-H335
    Highly flammable liquid and vapour.
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 126°C
    Quantity 100g
    Molecular Formula C6H10O2
    CAS 4454-05-1
    Beilstein 107274
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
    RTECS Number UP7900000
    Flash Point 16°C (61°F)
    Chemical Name or Material 3,4-Dihydro-2-methoxy-2H-pyran
    Assay 99%

    GHS H Statement
    H225-H301-H315-H319-H335
    Highly flammable liquid and vapour.
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit