Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2-methoxy-2H-pyran, +99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass Bottle
4454-05-1
C6H10O2
114.14
MFCD00006560
XCYWUZHUTJDTGS-UHFFFAOYNA-N
zinc ii gluconate hydrate,zinc 2+ bis d-gluconate hydrate
131674739
2-methoxy-3,4-dihydro-2H-pyran
COC1CCC=CO1
4454-05-1
C6H10O2
114.14
MFCD00006560
XCYWUZHUTJDTGS-UHFFFAOYNA-N
zinc ii gluconate hydrate,zinc 2+ bis d-gluconate hydrate
131674739
2-methoxy-3,4-dihydro-2H-pyran
COC1CCC=CO1
Water 0.2% max.
Flash Point 16°C
Refractive Index 1.4410 to 1.4430
Quantity 5g
Packaging Glass Bottle
Specific Gravity 1
Boiling Point 126.0°C
Color Colorless to Yellow
Density 1.0000g/mL
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Formula Weight 114.14
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material 3, 4-Dihydro-2-methoxy-2H-pyran, 99+%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Toxic if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-698-3

RTECSNumber : UP7900000

TSCA : TSCA