Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2-methoxy-2H-pyran 97.0+%, TCI America™
SDP

Catalog No. p-7126786
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 4454-05-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00006560
InChI Key XCYWUZHUTJDTGS-UHFFFAOYNA-N
Synonym 3,4-dihydro-2-methoxy-2h-pyran, 2h-pyran, 3,4-dihydro-2-methoxy, 2,3-dihydro-2-methoxy 4h pyran, 2-methoxy-2,3-dihydro-4h-pyran, acmc-1ahgh, dsstox_cid_7570, dsstox_rid_78508, dsstox_gsid_27570, ksc496o2j
PubChem CID 91538
IUPAC Name 2-methoxy-3,4-dihydro-2H-pyran
SMILES COC1CCC=CO1

Specifications

Color Yellow
Boiling Point 127°C
Quantity 25 mL
UN Number 1993
Formula Weight 114.14
Percent Purity ≥97.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material 3,4-Dihydro-2-methoxy-2H-pyran
EINECSNumber (5)-5581
RTECSNumber UP7900000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.