Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3,4-Dihydro-2H-1,5-benzodioxepin, 98%

Manufacturer:  Alfa Aesar™ H2737403

Quantity 1g

 View more versions of this product

Catalog No. AAH2737403


  Check Availability
Add to cart

Specifications

Description

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

7216-18-4
150.18
1,2-Trimethylenedioxybenzene
C9H10O2
MFCD01333457
3,4-Dihydro-2H-1,5-benzodioxepin

Specifications

1g
C9H10O2
MFCD01333457
3,4-Dihydro-2H-1,5-benzodioxepin
7216-18-4
150.18
1,2-Trimethylenedioxybenzene
98%

Safety and Handling

SDS & Safety Documents

RTECSNumber : DF4725200

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'